missing translation for 'onlineSavingsMsg'
Learn More

Carboxymethylcellulose Sodium, Medium Viscosity, USP, Spectrum™ Chemical

Catalog No. 18601481
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601481 125 g Poly Bottle
18601482 500 g Poly Bottle
18601483 2.5 kg Poly Pail
18601484 12 kg Poly Pail
18601485 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18601481 Supplier Spectrum Chemical Mfg Cor Supplier No. CA192125GM

CA192, 9004-32-4

Spectrum™ Chemical Carboxymethylcellulose Sodium, Medium Viscosity, Viscosity of 2 Percent Aqueous Solution @ 25 DEG C: 400-800 cP, USP is often used as a thickener and stabilizer. The USP grade indicates it is a grade suitable for personal care, cosmetic and pharmaceutical applications and were manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-32-4
Molecular Formula (C12 H14 O9 R6)n
Molecular Weight (g/mol) 263.20
MDL Number MFCD00081472
InChI Key DPXJVFZANSGRMM-UHFFFAOYNA-N
IUPAC Name 2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
SMILES [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O

Specifications

CAS Max % 100%
Quantity 125 g
Grade USP
Loss on Drying 10%
Packaging Poly Bottle