missing translation for 'onlineSavingsMsg'
Learn More

Carboxymethylcellulose Calcium Salt, Spectrum™ Chemical

Catalog No. 18601468
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601468 500 g Poly Bottle
18601466 25 g Poly Bottle
18601467 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18601468 Supplier Spectrum Chemical Mfg Cor Supplier No. C1528500GM
Only null left

C1528, 9050-04-8

Spectrum™ Chemical Carboxymethylcellulose Calcium Salt , also known as Carboxymethyl cellulose or cellulose gum, can be used as a thickening agent in pharmaceuticals. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 9050-04-8

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 500 g
Grade Reagent