missing translation for 'onlineSavingsMsg'
Learn More

Carboxymethylcellulose Calcium Salt, NF, Spectrum™ Chemical

C1721, 9050-04-8

Supplier:  Spectrum Chemical Mfg Cor C172110KGBL

Quantity 10 kg
Packaging Fiber Drum

 View more versions of this product

Catalog No. 18601470

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Spectrum™ Chemical Carboxymethylcellulose Calcium Salt, NF
Specifications

Chemical Identifiers

9050-4-8

Specifications

9050-4-8
10.0-20%
Fiber Drum
NF
100%
10%
10 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions