missing translation for 'onlineSavingsMsg'
Learn More

Carboplatin

Catalog No. p-7058185
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6043303 1 g
AAJ6043306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6043303 Supplier Thermo Scientific Chemicals Supplier No. J6043303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 41575-94-4 | C6H12N2O4Pt | 371.26 g/mol

Carboplatin is primarily used as an antitumor agent.Carboplatin is widely utilized as a chemotherapy drug to treat various types of cancer. It belongs to the group of platinum-based antineoplastic agents to interfere in DNA (deoxyribonucleic acid) repair. It is applied in combination other medications and used to inhibit cancer cell growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 41575-94-4
Molecular Formula C6H12N2O4Pt
Molecular Weight (g/mol) 371.26
MDL Number MFCD00070464
InChI Key BHKICZDKIIDMNR-UHFFFAOYSA-L
Synonym carboplatin
PubChem CID 122130863
IUPAC Name azanide;cyclobutane-1,1-dicarboxylate;platinum(4+)
SMILES N.N.O=C1O[Pt++]OC(=O)C11CCC1

Specifications

Color White
Linear Formula [C4H6(CO2)2]Pt(NH3)2
Quantity 1 g
Merck Index 14,1822
Solubility Information Soluble in water.
Formula Weight 371.25
Physical Form Powder
Chemical Name or Material Carboplatin
Hazard Category H302+H312+H332-H317-H334-H340-H350-H360
Hazard Statement GHS H Statement
H334-H340-H360-H302-H312-H332-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May damage fertility or the unborn child.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P333+P313-P342+P311-P363-P501c
EINECSNumber 255-446-0
RTECSNumber TP2300000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only