accessibility menu, dialog, popup

UserName

CAS RN 41575-94-4

IUPAC Name: 5,9-dioxo-6,8-dioxa-7-platinaspiro[3.5]nonane-7,7-bis(ylium) diamine
Synonyms: 5,9-dioxo-6,8-dioxa-7-platinaspiro[3.5]nonane-7,7-bis(ylium) diamine
Molecular Weight (g/mol): 371.26
Molecular Formula: C6H12N2O4Pt
InChi Key: BHKICZDKIIDMNR-UHFFFAOYSA-L
SMILES: N.N.O=C1O[Pt++]OC(=O)C11CCC1
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)