missing translation for 'onlineSavingsMsg'
Learn More

Carboplatin 98.0+%, TCI America™

Catalog No. C20431G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20431G 1 g
C2043100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20431G Supplier TCI America Supplier No. C20431G

Chemical Identifiers

CAS 41575-94-4
Molecular Formula C6H12N2O4Pt
Molecular Weight (g/mol) 371.26
MDL Number MFCD00070464
InChI Key BHKICZDKIIDMNR-UHFFFAOYSA-L
Synonym carboplatin
PubChem CID 122130863
IUPAC Name 5,9-dioxo-6,8-dioxa-7-platinaspiro[3.5]nonane-7,7-bis(ylium) diamine
SMILES N.N.O=C1O[Pt++]OC(=O)C11CCC1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 371.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Carboplatin
RTECSNumber TP2300000
TSCA No