missing translation for 'onlineSavingsMsg'
Learn More

Carbonylhydridotris(triphenylphosphine)iridium(I), TCI America™

Catalog No. C3040200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3040200MG 200 mg
C30401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3040200MG Supplier TCI America Supplier No. C3040200MG
Only null left

Chemical Identifiers

CAS 17250-25-8
Molecular Formula C55H46IrOP3
Molecular Weight (g/mol) 1008.11
MDL Number MFCD00015525
InChI Key IOPVMNYABUEALT-UHFFFAOYSA-N
Synonym carbonylhydridotris triphenylphosphine iridium i, iridium 1+ formaldehyde tris triphenylphosphine hydride
PubChem CID 92135237
IUPAC Name methanidylidyneoxidanium tris(triphenylphosphane) iridium hydride
SMILES [IrH].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 170°C
Color Yellow
Quantity 200 mg
UN Number 2811
Formula Weight 1008.11
Physical Form Crystalline Powder
Chemical Name or Material Carbonylhydridotris(triphenylphosphine)iridium(I)