missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonylchlorobis(triphenylphosphine)iridium(I) 95.0+%, TCI America™
$208.78 - $738.16
Chemical Identifiers
| CAS | 14871-41-1 |
|---|---|
| Molecular Formula | C37H30ClIrOP2 |
| Molecular Weight (g/mol) | 780.26 |
| MDL Number | MFCD00009590 |
| InChI Key | PEPSDKMVHYXIMJ-UHFFFAOYSA-M |
| Synonym | carbonylchlorobis triphenylphosphine iridium i |
| PubChem CID | 78076234 |
| IUPAC Name | chloroiridiumylium; methanidylidyneoxidanium; bis(triphenylphosphane) |
| SMILES | Cl[Ir+].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 14871-41-1 | |
| 780.26 | |
| PEPSDKMVHYXIMJ-UHFFFAOYSA-M | |
| 78076234 | |
| Cl[Ir+].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
| C37H30ClIrOP2 | |
| MFCD00009590 | |
| carbonylchlorobis triphenylphosphine iridium i | |
| chloroiridiumylium; methanidylidyneoxidanium; bis(triphenylphosphane) |
Specifications
| 14871-41-1 | |
| Yellow | |
| MFCD00009590 | |
| 2811 | |
| PEPSDKMVHYXIMJ-UHFFFAOYSA-M | |
| chloroiridiumylium; methanidylidyneoxidanium; bis(triphenylphosphane) | |
| 78076234 | |
| ≥95.0% (T) | |
| Carbonylchlorobis(triphenylphosphine)iridium(I) |
| 225°C | |
| C37H30ClIrOP2 | |
| 200 mg | |
| carbonylchlorobis triphenylphosphine iridium i | |
| Cl[Ir+].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 780.26 | |
| 780.26 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes