missing translation for 'onlineSavingsMsg'
Learn More

Carbonyl(dihydrido)tris(triphenylphosphine)ruthenium(II), TCI America™

Catalog No. p-7125361
Change view
Click to view available options
Quantity:
250 mg
1 g
Catalog No. Quantity
C2251250MG 250 mg
C22511G 1 g
2 options

Catalog No. C2251250MG

Supplier: TCI America C2251250MG

Only null left

Chemical Identifiers

CAS 25360-32-1
Molecular Formula C55H47OP3Ru
Molecular Weight (g/mol) 917.97
MDL Number MFCD00015870 MFCD00015870
InChI Key MLIYPCQSOXNTLJ-UHFFFAOYSA-N
PubChem CID 53384314
IUPAC Name methanidylidyneoxidanium tris(triphenylphosphane) λ²-ruthenium
SMILES [RuH2].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 163°C
Color White-Yellow
Quantity 250 mg
Formula Weight 917.97
Physical Form Crystalline Powder
Chemical Name or Material Carbonyl(dihydrido)tris(triphenylphosphine)ruthenium(II)