missing translation for 'onlineSavingsMsg'
Learn More

Carbonyl cyanide 3-chlorophenylhydrazone, 98%

Catalog No. aal06932mc
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL06932MC 100 mg
AAL06932ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL06932MC Supplier Thermo Scientific Chemicals Supplier No. L06932MC
Only null left

CAS: 555-60-2 | C9H5ClN4 | 204.617 g/mol

Carbonyl cyanide 3-chlorophenylhydrazone is a widely used uncoupler of oxidative phosphorylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbonyl cyanide 3-chlorophenylhydrazone is a widely used uncoupler of oxidative phosphorylation.

Solubility
Insoluble in water. Soluble in DMSO (5 mg/ml), ethanol (1 mg/ml) and methanol (10 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 555-60-2
Molecular Formula C9H5ClN4
Molecular Weight (g/mol) 204.617
MDL Number MFCD00001848
InChI Key UGTJLJZQQFGTJD-UHFFFAOYSA-N
Synonym cccp, 3-chlorophenyl hydrazonomalononitrile, carbonylcyanide-3-chlorophenylhydrazone, carbonyl cyanide 3-chlorophenylhydrazone, carbonyl cyanide m-chlorophenyl hydrazone, carbonyl cyanide m-chlorophenylhydrazone, 3-chlorophenyl hydrazono malononitrile, m-chlorophenyl carbonylcyanide hydrazone, m-cl-ccp, carbonyl cyanide, 3-chlorophenyl hydrazone
PubChem CID 2603
ChEBI CHEBI:3259
IUPAC Name 2-[(3-chlorophenyl)hydrazinylidene]propanedinitrile
SMILES C1=CC(=CC(=C1)Cl)NN=C(C#N)C#N

Specifications

Melting Point ∽175°C (decomposition)
Odor Odorless
Quantity 100 mg
UN Number UN3439
Beilstein 1842102
Solubility Information Insoluble in water. Soluble in DMSO (5mg/ml),ethanol (1mg/ml) and methanol (10mg/ml).
Formula Weight 204.62
Percent Purity 98%
Chemical Name or Material Carbonyl cyanide 3-chlorophenylhydrazone
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 209-103-7
RTECSNumber FG5600000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only