Learn More
Carbonyl cyanide 3-chlorophenylhydrazone, 98%
CAS: 555-60-2 | C9H5ClN4 | 204.62 g/mol
Supplier: Thermo Scientific Chemicals 228130010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Carbonyl cyanide 3-chlorophenylhydrazone | |
| 555-60-2 | |
| Authentic | |
| Glass bottle | |
| ClC6H4NHN=C(CN)2 | |
| 1 g | |
| Solubility in water: insoluble | |
| C1=CC(=CC(=C1)Cl)NN=C(C#N)C#N | |
| 204.62 | |
| CHEBI:3259 | |
| 98% |
| 99+% | |
| 172°C to 177°C | |
| 97.5% min. (HPLC) | |
| C9H5ClN4 | |
| MFCD00001848 | |
| cccp, 3-chlorophenyl hydrazonomalononitrile, carbonylcyanide-3-chlorophenylhydrazone, carbonyl cyanide 3-chlorophenylhydrazone, carbonyl cyanide m-chlorophenyl hydrazone, carbonyl cyanide m-chlorophenylhydrazone, 3-chlorophenyl hydrazono malononitrile, m-chlorophenyl carbonylcyanide hydrazone, m-cl-ccp, carbonyl cyanide, 3-chlorophenyl hydrazone | |
| UGTJLJZQQFGTJD-UHFFFAOYSA-N | |
| 2-[(3-chlorophenyl)hydrazinylidene]propanedinitrile | |
| 2603 | |
| 204.62 | |
| Crystalline Powder |
Chemical Identifiers
| 555-60-2 | |
| 204.62 | |
| UGTJLJZQQFGTJD-UHFFFAOYSA-N | |
| 2603 | |
| 2-[(3-chlorophenyl)hydrazinylidene]propanedinitrile |
| C9H5ClN4 | |
| MFCD00001848 | |
| cccp, 3-chlorophenyl hydrazonomalononitrile, carbonylcyanide-3-chlorophenylhydrazone, carbonyl cyanide 3-chlorophenylhydrazone, carbonyl cyanide m-chlorophenyl hydrazone, carbonyl cyanide m-chlorophenylhydrazone, 3-chlorophenyl hydrazono malononitrile, m-chlorophenyl carbonylcyanide hydrazone, m-cl-ccp, carbonyl cyanide, 3-chlorophenyl hydrazone | |
| CHEBI:3259 | |
| C1=CC(=CC(=C1)Cl)NN=C(C#N)C#N |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
GHS Signal Word: Danger
EINECSNumber : 209-103-7
RTECSNumber : FG5600000
TSCA : TSCA
RUO – Research Use Only