missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase VA Monoclonal Antibody (K52035_18D12), Invitrogen™
Mouse Monoclonal Antibody
$1421.98 - $2134.69
Specifications
| Antigen | Carbonic Anhydrase VA |
|---|---|
| Clone | K52035_18D12 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: WQTSNNTLHP LWTVPVSVPG GTRQSPINIQ WRDSVYDPQL KPLRVSYEAA SCLYIWNTGY LFQVEFDDAT EASGISGGPL ENHYRLKQFH FHWGAVNEGG SEHTVDGHAY PAELHLVHWN SVKYQNYKEA VVGENGLAVI GVFLKLGAHH QTLQRLVDIL PEIKHKDARA AMRPFDPSTL LPTCWDYWTY AGSLTTPPLT ESVTWIIQKE PVEVAPSQLS AFRTLLFSAL GEEEKMMVNN YRPLQPLMNR KVWASFQATN
Carbonic Anhydrase VA Monoclonal antibody specifically detects Carbonic Anhydrase VA in Human, Rat samples. It is validated for Western BlotSpecifications
| Carbonic Anhydrase VA | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| CA Y; CA5; CA5A; CA5AD; Car5; Car5a; Carbonate dehydratase VA; carbonic anhydrase 5 mitochondrial; carbonic anhydrase 5, mitochondrial; carbonic anhydrase 5A; carbonic anhydrase 5a, mitochondrial; carbonic anhydrase 5b, mitochondrial; carbonic anhydrase V, mitochondrial; carbonic anhydrase VA; carbonic anhydrase VA, mitochondrial; carbonic dehydratase; CAV; CAVA; CA-VA; GS1-21A4.1 | |
| Recombinant human CA5A (immunogen range is 41-300). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| CA5A |
| K52035_18D12 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| P35218, P43165 | |
| 54233, 763 | |
| IgG1 | |
| Protein A | |
| Antibody |