missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Carbonic Anhydrase VA Monoclonal Antibody (K52035_18D12), Invitrogen™

Catalog No. MA555641
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555641 50 μL
MA556021 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555641 Supplier Invitrogen™ Supplier No. MA555641
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: WQTSNNTLHP LWTVPVSVPG GTRQSPINIQ WRDSVYDPQL KPLRVSYEAA SCLYIWNTGY LFQVEFDDAT EASGISGGPL ENHYRLKQFH FHWGAVNEGG SEHTVDGHAY PAELHLVHWN SVKYQNYKEA VVGENGLAVI GVFLKLGAHH QTLQRLVDIL PEIKHKDARA AMRPFDPSTL LPTCWDYWTY AGSLTTPPLT ESVTWIIQKE PVEVAPSQLS AFRTLLFSAL GEEEKMMVNN YRPLQPLMNR KVWASFQATN

Carbonic anhydrases (CAs) are members of a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. CAs are involved in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric juice. They show extensive diversity in distribution and in their subcellular localization. The human CA5A gene encodes for the CA V protein which is expressed in the mitochondrial matrix in liver tissues, but can also be detected in many other tissues. In ureagenesis, the CA V protein may be involved in providing bicarbonate to carbamoyl phosphate synthetase.

Specifications

Antigen Carbonic Anhydrase VA
Applications Western Blot
Classification Monoclonal
Clone K52035_18D12
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene CA5A
Gene Accession No. P35218, P43165
Gene Alias CA Y; CA5; CA5A; CA5AD; Car5; Car5a; Carbonate dehydratase VA; carbonic anhydrase 5 mitochondrial; carbonic anhydrase 5, mitochondrial; carbonic anhydrase 5A; carbonic anhydrase 5a, mitochondrial; carbonic anhydrase 5b, mitochondrial; carbonic anhydrase V, mitochondrial; carbonic anhydrase VA; carbonic anhydrase VA, mitochondrial; carbonic dehydratase; CAV; CAVA; CA-VA; GS1-21A4.1
Gene Symbols CA5A
Host Species Mouse
Immunogen Recombinant human CA5A (immunogen range is 41-300).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54233, 763
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less