missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Carbonate, 0.2M buffer soln., pH 9.6, low endotoxin

$108.29 - $148.85

Specifications

Color Colorless
Concentration 0.2 M buffer soln.
pH Range 9.6
Format Liquid
Storage Requirements Ambient temperatures
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ67254AE
View Documents Promotion Details
Thermo Scientific™
J67254AE
100 mL
Each for $108.29
Only null left
 
AAJ67254AK
View Documents Promotion Details
Thermo Scientific™
J67254AK
250 mL
Each for $148.85
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Colorless
9.6
Ambient temperatures
0.2 M buffer soln.
Liquid
Carbonate
Videos
SDS
Documents

Documents