missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Carbonate, 0.2M buffer soln., pH 9.6, low endotoxin

Catalog No. AAJ67254AK
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67254AK 250 mL
AAJ67254AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67254AK Supplier Thermo Scientific™ Supplier No. J67254AK

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Color Colorless
Concentration 0.2 M buffer soln.
pH Range 9.6
Format Liquid
Quantity 250 mL
Storage Requirements Ambient temperatures
Type Carbonate