missing translation for 'onlineSavingsMsg'
Learn More

Carbon Tetraiodide 97.0+%, TCI America™

Catalog No. C093610G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C093610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C093610G Supplier TCI America Supplier No. C093610G

Chemical Identifiers

CAS 507-25-5
Molecular Formula CI4
Molecular Weight (g/mol) 519.629
MDL Number MFCD00001067
InChI Key JOHCVVJGGSABQY-UHFFFAOYSA-N
Synonym Tetraiodomethane
PubChem CID 10487
IUPAC Name tetraiodomethane
SMILES C(I)(I)(I)I

Specifications

Melting Point 171°C
Color Black-Gray
Quantity 10 g
Formula Weight 519.63
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Carbon Tetraiodide
RTECSNumber FG4960000
TSCA Yes
Recommended Storage Refrigerator