accessibility menu, dialog, popup

UserName

IIII

CAS RN 507-25-5

IUPAC Name: tetraiodomethane
Synonyms: carbon tetraiodide
Molecular Weight (g/mol): 519.63
Molecular Formula: CI4
InChi Key: JOHCVVJGGSABQY-UHFFFAOYSA-N
SMILES: IC(I)(I)I
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results