missing translation for 'onlineSavingsMsg'
Learn More

Carbon Tetrabromide 99.0+%, TCI America™

Catalog No. p-7136046
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T003825G 25 g
T0038100G 100 g
T0038500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T003825G Supplier TCI America Supplier No. T003825G
Only null left

Chemical Identifiers

CAS 558-13-4
Molecular Formula CBr4
Molecular Weight (g/mol) 331.627
MDL Number MFCD00000117
InChI Key HJUGFYREWKUQJT-UHFFFAOYSA-N
Synonym carbon tetrabromide, methane, tetrabromo, carbon bromide, methane tetrabromide, bromid uhlicity, carbontetrabromide, methane, tetrabromide, tetrabrommethan, bromid uhlicity czech
PubChem CID 11205
ChEBI CHEBI:47875
IUPAC Name tetrabromomethane
SMILES C(Br)(Br)(Br)Br

Specifications

Melting Point 94°C
Color White
Boiling Point 190°C
Quantity 25 g
UN Number 2516
Formula Weight 331.63
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Carbon Tetrabromide
EINECSNumber (2)-0041
RTECSNumber FG4725000
TSCA Yes