Learn More
Carbon tetrabromide, 98% (dry wt.), may cont. up to ca 6% water
CAS: 558-13-4 | CBr4 | 331.627 g/mol
Supplier: Thermo Scientific Chemicals L0082136
Description
Carbon tetrabromide is widely utilized as a solvent for greases, waxes and oils. In Appel reaction, it is used to convert alcohols to alkyl bromides. It has the property of non-flammability and finds application in fire-resistant chemicals as an active ingredient. It is also used for separating minerals. It can serves as an intermediate in manufacturing agrochemicals. It plays a vital role in the plastic and rubber industries for blowing and vulcanization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCarbon tetrabromide is widely utilized as a solvent for greases, waxes and oils. In Appel reaction, it is used to convert alcohols to alkyl bromides. It has the property of non-flammability and finds application in fire-resistant chemicals as an active ingredient. It is also used for separating minerals. It can serves as an intermediate in manufacturing agrochemicals. It plays a vital role in the plastic and rubber industries for blowing and vulcanization.
Solubility
Insoluble in water. Soluble in ether, chloroform and ethanol.
Notes
Store in cool place. Incompatible with strong oxidizing agents, strong bases and lithium.
Specifications
| Carbon tetrabromide | |
| 90°C to 95°C | |
| 190°C | |
| CBr4 | |
| 500 g | |
| 1732799 | |
| Insoluble in water. Soluble in ether,chloroform and ethanol. | |
| C(Br)(Br)(Br)Br | |
| 331.627 | |
| CHEBI:47875 | |
| 98% |
| 558-13-4 | |
| 3.42 | |
| (dry wt.), may cont. up to ≈6% water | |
| MFCD00000117 | |
| UN2516 | |
| carbon tetrabromide, methane, tetrabromo, carbon bromide, methane tetrabromide, bromid uhlicity, carbontetrabromide, methane, tetrabromide, tetrabrommethan, bromid uhlicity czech | |
| HJUGFYREWKUQJT-UHFFFAOYSA-N | |
| tetrabromomethane | |
| 11205 | |
| 331.63 |
Chemical Identifiers
| 558-13-4 | |
| 331.627 | |
| HJUGFYREWKUQJT-UHFFFAOYSA-N | |
| 11205 | |
| tetrabromomethane |
| CBr4 | |
| MFCD00000117 | |
| carbon tetrabromide, methane, tetrabromo, carbon bromide, methane tetrabromide, bromid uhlicity, carbontetrabromide, methane, tetrabromide, tetrabrommethan, bromid uhlicity czech | |
| CHEBI:47875 | |
| C(Br)(Br)(Br)Br |
Safety and Handling
GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CARBON TETRABROMIDE
EINECSNumber : 209-189-6
RTECSNumber : FG4725000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only