missing translation for 'onlineSavingsMsg'
Learn More

Carbon Standard, Organic, 200 ppm, Reagents

Catalog No. 01335592
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335592 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335592 Supplier Labchem Inc Supplier No. CC2054001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Carbon Standard, Organic, 200 ppm, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Carbon Standard, Organic, 200 ppm
Packaging Glass Bottle
CAS 7732-18-5, 877-24-7
Specific Gravity 1.0002
Physical Form Liquid
Color Colorless
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Industry Type Water Analysis
Quantity 1 L
Grade Reagent
Show More Show Less