missing translation for 'onlineSavingsMsg'
Learn More

Carbon Standard For ICP, 1000μg/mL in H2O, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLC92X

 View more versions of this product

Catalog No. PLC9-2X


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
C1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Carbon Standard
7440-44-0,7732-18-5
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
0.1,99.9
AA, ICP
Certified Reference Material
[C]
carbon
500 mL
CHEBI:27594
Matrix Concentration Cat. No.
Oxalic Acid in H2O 1000μg/mL PLC9-2Y, -2X 
Triple-leached LDPE bottle
Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
0.1,99.9
C
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
OKTJSMMVPCPJKN-UHFFFAOYSA-N
Water
12.011
5462310
Assurance

Chemical Identifiers

7440-44-0
12.011
5462310
carbon
C
OKTJSMMVPCPJKN-UHFFFAOYSA-N
CHEBI:27594
[C]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only