Learn More
Carbon powder, activated, Norit GSX, steam activated, acid washed
CAS: 7440-44-0 | C | 12.011 g/mol
Supplier: Thermo Scientific Chemicals L118600P
Description
Decolorizing carbon. High purity grade with high internal surface area, suitable for decolorization, purification and catalyst carrier applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Carbon powder | |
| 7440-44-0 | |
| MFCD00133992 | |
| UN1362 | |
| OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
| carbon | |
| 5462310 |
| activated, Norit GSX, steam activated, acid washed | |
| C | |
| 2000 g | |
| graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
| [C] | |
| 12.011 | |
| CHEBI:27594 |
Chemical Identifiers
| 7440-44-0 | |
| 12.011 | |
| OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
| 5462310 | |
| carbon |
| C | |
| MFCD00133992 | |
| graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
| CHEBI:27594 | |
| [C] |
Safety and Handling
GHS H Statement
H228-H252
Flammable solid.
Self-heating in large quantities; may catch fire.
P235+P410-P280
H252
DOTInformation : Transport Hazard Class: 4.2; Packing Group: III; Proper Shipping Name: CARBON, ACTIVATED
EINECSNumber : 231-153-3
RTECSNumber : FF5250100
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only