missing translation for 'onlineSavingsMsg'
Learn More

Carbon powder, activated, Norit GSX, steam activated, acid washed

Catalog No. p-7045754
Change view
Click to view available options
Quantity:
100 g
500 g
2000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1186022 100 g
AAL1186036 500 g
AAL118600P 2000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1186022 Supplier Thermo Scientific Chemicals Supplier No. L1186022
Only null left

CAS: 7440-44-0 | C | 12.011 g/mol

Decolorizing carbon. High purity grade with high internal surface area, suitable for decolorization, purification and catalyst carrier applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Quantity 100 g
UN Number UN1362
Chemical Name or Material Carbon powder, activated, Norit GSX, steam activated, acid washed
Hazard Category H252
Hazard Statement GHS H Statement
H228-H252
Flammable solid.
Self-heating in large quantities; may catch fire.
Precautionary Statement P235+P410-P280
DOTInformation Transport Hazard Class: 4.2; Packing Group: III; Proper Shipping Name: CARBON, ACTIVATED
EINECSNumber 231-153-3
RTECSNumber FF5250100
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only