missing translation for 'onlineSavingsMsg'
Learn More

Carbon Disulfide (Low Benzene), Fisher Chemical

Catalog No. c185500
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C185500 500 mL Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C185500 Supplier Thermo Fisher Scientific Supplier No. FLC185500
Only null left

Chemical Identifiers

CAS 75-15-0
Molecular Formula CS2
Molecular Weight (g/mol) 76.131
MDL Number MFCD00011321
InChI Key QGJOPFRUJISHPQ-UHFFFAOYSA-N
Synonym carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
PubChem CID 6348
ChEBI CHEBI:23012
IUPAC Name methanedithione
SMILES C(=S)=S

Specifications

Melting Point -111°C
Absorbance 0.01 AU max. at 500nm, 0.05 AU max. at 410nm, 0.10 AU max. at 403nm, 0.50 AU max. at 388nm, 1.0 AU max. at 382nm
Color Yellow
Quantity 500 mL
Identification Pass Test
Packaging Amber Glass
Physical Form Liquid
Residue after Evaporation 0.002% max.
Assay Percent Range ≥99.9 %
Vapor Pressure 297.5mmHg at 20°C
Viscosity 0.363 cps at 20 deg C
Water 0.05% max.
Boiling Point 46°C
Formula Weight 76.14
Percent Purity ≥99.9%
Chemical Name or Material Carbon Disulfide, Low Benzene
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:3
Flammability:4
Instability:0