Learn More
Carbon disulfide, HPLC Grade, 99.8+%
CAS: 75-15-0 | CS2 | 76.131 g/mol
$229.49 - $229.49
Chemical Identifiers
| CAS | 75-15-0 |
|---|---|
| Molecular Formula | CS2 |
| Molecular Weight (g/mol) | 76.131 |
| MDL Number | MFCD00011321 |
| InChI Key | QGJOPFRUJISHPQ-UHFFFAOYSA-N |
| Synonym | carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide |
| PubChem CID | 6348 |
| ChEBI | CHEBI:23012 |
| IUPAC Name | methanedithione |
| SMILES | C(=S)=S |
Description
Carbon disulfide is used in spectrophotometry, high performance liquid chromatography and environmental testing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 75-15-0 | |
| 76.131 | |
| QGJOPFRUJISHPQ-UHFFFAOYSA-N | |
| 6348 | |
| methanedithione |
| CS2 | |
| MFCD00011321 | |
| carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide | |
| CHEBI:23012 | |
| C(=S)=S |
Specifications
| -112°C | |
| 1.2632 g/mL | |
| 1 L | |
| Unpleasant | |
| 1.627 | |
| CS2 | |
| UN1131 | |
| carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide | |
| QGJOPFRUJISHPQ-UHFFFAOYSA-N | |
| methanedithione | |
| 6348 | |
| 76.14 | |
| HPLC |
| 75-15-0 | |
| 46°C | |
| -30°C (-22°F) | |
| Liquid | |
| >99.8% | |
| MFCD00011321 | |
| 14,1811 | |
| Miscible with alcohol,ether,benzene,oil,chloroform and carbon tetrachloride. | |
| C(=S)=S | |
| 76.131 | |
| CHEBI:23012 | |
| ≥99.8% | |
| Carbon disulfide |
Safety and Handling
GHS H Statement
H225-H372-H361-H302-H315-H319 Highly flammable liquid and vapor.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H332-H336-H361fd-H372
DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: CARBON DISULFIDE
EINECSNumber : 200-843-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only