missing translation for 'onlineSavingsMsg'
Learn More

Carbon Disulfide (Certified ACS), Fisher Chemical™

Catalog No. p-215806
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Amber Glass Bottle
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C184500 500 mL Amber Glass Bottle
C184212 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C184500 Supplier Thermo Fisher Scientific Supplier No. FLC184500
Only null left

Chemical Identifiers

CAS 75-15-0
Molecular Formula CS2
Molecular Weight (g/mol) 76.131
MDL Number MFCD00011321
InChI Key QGJOPFRUJISHPQ-UHFFFAOYSA-N
Synonym carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
PubChem CID 6348
ChEBI CHEBI:23012
IUPAC Name methanedithione
SMILES C(=S)=S

Specifications

Melting Point -111°C
Color Yellow
Quantity 500 mL
Flash Point 35°C (95°F)
Packaging Amber Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.002% max.
Assay Percent Range ≥99.9%
Vapor Pressure 297.5mmHg at 20°C
Viscosity 0.363 cps at 20 deg C
UN Number 1120; Colorless
Water 0.05% max.
Boiling Point 46°C
Formula Weight 76.14
Percent Purity ≥99.9%
Grade ACS
Chemical Name or Material Carbon Disulfide
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:3
Flammability:4
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid