missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon disulfide, ACS Reagent, ≥99.9%, Honeywell Riedel-de Haën™
ACS Reagent,≥99.9%
$373.13 - $373.13
Chemical Identifiers
| CAS | 75-15-0 |
|---|---|
| Molecular Formula | CS2 |
| Molecular Weight (g/mol) | 76.131 |
| MDL Number | MFCD00011321 |
| InChI Key | QGJOPFRUJISHPQ-UHFFFAOYSA-N |
| Synonym | carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide |
| PubChem CID | 6348 |
| ChEBI | CHEBI:23012 |
| IUPAC Name | methanedithione |
| SMILES | C(=S)=S |
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Chemical Identifiers
| 75-15-0 | |
| 76.131 | |
| QGJOPFRUJISHPQ-UHFFFAOYSA-N | |
| 6348 | |
| methanedithione |
| CS2 | |
| MFCD00011321 | |
| carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide | |
| CHEBI:23012 | |
| C(=S)=S |
Specifications
| -76°C | |
| Colorless | |
| 46°C (1013 hPa) | |
| 0.5 | |
| Unpleasant | |
| Liquid | |
| MFCD00011321 | |
| 1098293 | |
| 2.9g/L in water at 20°C | |
| C(=S)=S | |
| 76.131 | |
| CHEBI:23012 | |
| ≥99.9% | |
| Carbon disulfide |
| 75-15-0 | |
| 1.266g/mL at 25°C | |
| 1 L | |
| 212°C | |
| Glass Bottle | |
| CS2 | |
| UN1131 | |
| carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide | |
| QGJOPFRUJISHPQ-UHFFFAOYSA-N | |
| methanedithione | |
| 6348 | |
| 76.14g/mol | |
| ACS Reagent |
Safety and Handling
P201-P210-P261-P280-P304 + P340 + P312-P308 + P313
DOTInformation : class 3 - PG 1 - Carbon disulfide
RTECSNumber : 200-843-6