missing translation for 'onlineSavingsMsg'
Learn More

Carbon Disulfide, ACS, 99.9%, Spectrum™ Chemical

$797.43 - $797.43

Chemical Identifiers

CAS 75-15-0
Molecular Weight (g/mol) 76.13
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18609574
View Documents
Spectrum Chemical Mfg Cor
C1170500MLGL
500 mL Amber Glass Bottle
Each for $797.43
Only null left
 
Description

Description

Specifications

Chemical Identifiers

75-15-0
76.13

Specifications

75-15-0
46.3°C
Amber Glass Bottle
76.13
Reagent
1
500 mL
2e-05
0.999
Carbon Disulfide
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.