Learn More
MilliporeSigma™ Supelco™ Carbon Dioxide Trap
As carbon dioxide is absorbed, the greenish-brown material turns white due to of the formation of sodium carbonate. The trap body is constructed of borosilicate glass, and the fittings are nickel-plated with sintered stainless steel frits.
Supplier:  MilliporeSigma™ Supelco™ 503193
 View more versions of this product
Catalog No. 111022613
Only null left
Explore available promotions
Description
Description
  • Sodium hydroxide nonfibrous silicate material is universally accepted for its high absorptive capacity and indicating properties for carbon dioxide
  • Typically, this material will absorb 20-30% of its weight in carbon dioxide before replacement of the saturated material is required
  • A moisture trap should be installed downstream from this unit to absorb the moisture that is produced
  • Exercise extreme caution when refilling the trap due to the caustic nature of the absorbent
Product Suggestions
Product Suggestions