missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon, activated, -4+8 mesh
CAS: 1333-86-4 | C | 12.01 g/mol
Supplier: Thermo Scientific Chemicals 043118A3
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Carbon, activated | |
| -4+8 Mesh | |
| MFCD00133992 | |
| graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
| [C] | |
| 12.01 | |
| CHEBI:27594 |
| 1333-86-4 | |
| C | |
| 2 kg | |
| OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
| carbon | |
| 5462310 |
Chemical Identifiers
| 1333-86-4 | |
| 12.01 | |
| OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
| 5462310 | |
| carbon |
| C | |
| MFCD00133992 | |
| graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
| CHEBI:27594 | |
| [C] |
Safety and Handling
H500
EINECSNumber : 231-153-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only