missing translation for 'onlineSavingsMsg'
Learn More

Carbon, activated, -4+8 mesh

Catalog No. p-7058721
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4311822 100 g
AA4311836 500 g
AA43118A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4311822 Supplier Thermo Scientific Chemicals Supplier No. 04311822
Only null left

CAS: 1333-86-4 | C | 12.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1333-86-4
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N

Specifications

Mesh Size −4+8 Mesh
Quantity 100 g
Chemical Name or Material Carbon, Activated
Hazard Category H500
EINECSNumber 231-153-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only