missing translation for 'onlineSavingsMsg'
Learn More

Carbon, activated, -4+8 mesh

CAS: 1333-86-4 | C | 12.01 g/mol

Supplier:  Thermo Scientific Chemicals 04311836

 View more versions of this product

Catalog No. AA4311836


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Carbon
1333-86-4
C
500 g
12.01
Activated
−4+8 Mesh
MFCD00133992
OKTJSMMVPCPJKN-UHFFFAOYSA-N

Chemical Identifiers

1333-86-4
12.01
OKTJSMMVPCPJKN-UHFFFAOYSA-N
C
MFCD00133992
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H500

EINECSNumber : 231-153-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only