Learn More
Carbomer 940, NF, Spectrum™ Chemical
C1477, 9003-01-4
Supplier:  Spectrum Chemical Mfg Cor C147725KG
 View more versions of this product
Catalog No. 18601456
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical Carbomer 940, NF is a polyvinyl carboxy polymer. It is used as a viscosity enhancer, gelling agent or suspension agent. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications and were manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
9003-01-4
(C3H4O2)n
2.5 kg
OC(=O)C(-*)C-*
72.06
2%
100%
MFCD00084394
NNMHYFLPFNGQFZ-UHFFFAOYSA-M
Poly(acrylic acid)
NF
Poly Pail
Chemical Identifiers
9003-01-4
72.06
NNMHYFLPFNGQFZ-UHFFFAOYSA-M
OC(=O)C(-*)C-*
(C3H4O2)n
MFCD00084394
Poly(acrylic acid)
Product Suggestions
Product Suggestions