missing translation for 'onlineSavingsMsg'
Learn More

Carbomer 940, NF, Spectrum™ Chemical

Catalog No. p-7157176
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601454 100 g Poly Bottle
18601455 500 g Poly Pail
18601456 2.5 kg Poly Pail
18601457 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601454 Supplier Spectrum Chemical Mfg Cor Supplier No. C1477100GM
Only null left

C1477, 9003-01-4

Spectrum™ Chemical Carbomer 940, NF is a polyvinyl carboxy polymer. It is used as a viscosity enhancer, gelling agent or suspension agent. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications and were manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9003-01-4
Molecular Formula (C3H4O2)n
Molecular Weight (g/mol) 72.06
MDL Number MFCD00084394
InChI Key NNMHYFLPFNGQFZ-UHFFFAOYSA-M
IUPAC Name Poly(acrylic acid)
SMILES OC(=O)C(-*)C-*

Specifications

CAS Max % 100%
Quantity 100 g
Grade NF
Loss on Drying 2%
Packaging Poly Bottle