missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Carbol Fuchsin, pure

Catalog No. AC195790050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC195790050 5 g Glass bottle
AC195790250 25 g Glass bottle
AC195791000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC195790050 Supplier Thermo Scientific Chemicals Supplier No. 195790050
Only null left

CAS: 4197-24-4 | C21H22ClN3 | 351.878 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4197-24-4
Molecular Formula C21H22ClN3
Molecular Weight (g/mol) 351.878
MDL Number MFCD00143923
InChI Key HZLHRDBTVSZCBS-GHTYLULLSA-N
PubChem CID 91997463
IUPAC Name 4-[(Z)-(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-2-methylaniline;hydron;chloride
SMILES [H+].CC1=CC(=C(C2=CC=C(C=C2)N)C3=CC(=C(C=C3)N)C)C=CC1=N.[Cl-]

Specifications

Infrared Spectrum Authentic
Lambda Max (in water) about 542nm
Grade Pure
Packaging Glass bottle
Suitability (for microscopy) passes
UV Spectrometry Complies
Color Green
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Carbol Fuchsin, A mixture of Basic Fuchsin and phenol.
Hazard Category Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical attention/advice.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
DOTInformation DOT Class 6.1
EINECSNumber 224-086-6

RUO – Research Use Only