accessibility menu, dialog, popup

UserName

CAS RN 4197-24-4

HClCH3H2NNH2NHH3C(Z)

CAS RN 4197-24-4

IUPAC Name: hydrogen 4-[(4-aminophenyl)[(1Z)-4-imino-3-methylcyclohexa-2,5-dien-1-ylidene]methyl]-2-methylaniline chloride
Synonyms: hydrogen 4-[(4-aminophenyl)[(1Z)-4-imino-3-methylcyclohexa-2,5-dien-1-ylidene]methyl]-2-methylaniline chloride
Molecular Weight (g/mol): 351.88
Molecular Formula: C21H22ClN3
InChi Key: HZLHRDBTVSZCBS-GHTYLULLSA-N
SMILES: [H+].[Cl-].CC1=CC(=CC=C1N)C(\C1=CC=C(N)C=C1)=C1\C=CC(=N)C(C)=C1
Color 
  • (3)
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)