missing translation for 'onlineSavingsMsg'
Learn More

Medisca Carbidopa, USP

Catalog No. 14185443
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185443 5 g
14185108 100 g
14185425 25 g
14185395 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14185443 Supplier Medisca Supplier No. 020203

Carbidopa USP is a white to creamy white, odorless or practically odorless, powder. Alternative name(s): carbidopa monohydrate

Carbidopa USP is a white to creamy white, odorless or practically odorless, powder.

Alternative name(s): carbidopa monohydrate

Specifications

Chemical Name or Material Carbidopa, USP
Packaging Bottle, 30mL, Amber Glass, Wide Mouth Packer, 28-400
CAS 38821-49-7
Type Drug Standard
Molecular Weight (g/mol) 244.245
Quantity 5 g
Percent Purity 98 to 102 %
Grade USP