missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Carbidopa, USP
Description
Carbidopa USP is a white to creamy white, odorless or practically odorless, powder.
Alternative name(s): carbidopa monohydrate
Specifications
Specifications
| Chemical Name or Material | Carbidopa, USP |
| Packaging | Jar, 300mL, White, HDPE, Round, 63-400, TE Pano Neck |
| CAS | 38821-49-7 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 244.245 |
| Quantity | 100 g |
| Percent Purity | 98 to 102 % |
| Grade | USP |