missing translation for 'onlineSavingsMsg'
Learn More

Carbetapentane citrate

Catalog No. AAJ64446MF
Change view
Click to view available options
Quantity:
50 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64446MF 50 mg
AAJ6444603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64446MF Supplier Thermo Scientific Chemicals Supplier No. J64446MF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sigma receptor agonist | CAS: 23142-01-0 | C26H39NO10 | 525.595 g/mol

Sigma receptor agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23142-01-0
Molecular Formula C26H39NO10
Molecular Weight (g/mol) 525.595
MDL Number MFCD00055697
InChI Key AKJDEXBCRLOVTH-UHFFFAOYSA-N
Synonym carbetapentane citrate, pentoxyverine citrate, loucarbate, pentoxiverine citrate, carbetapentane citrate salt, toclase, toclase citrate, carbetapentone citrate, unii-4sh0mfj5hj
PubChem CID 90010
IUPAC Name 2-[2-(diethylamino)ethoxy]ethyl 1-phenylcyclopentane-1-carboxylate;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES CCN(CC)CCOCCOC(=O)C1(CCCC1)C2=CC=CC=C2.C(C(=O)O)C(CC(=O)O)(C(=O)O)O

Specifications

Color White
Quantity 50 mg
Merck Index 14,1794
Formula Weight 633.7
Physical Form Powder
Chemical Name or Material Carbetapentane citrate
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 245-449-5
RTECSNumber GY3500000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only