missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbenicillin disodium salt, MP Biomedicals™
$182.39 - $626.88
Chemical Identifiers
| CAS | 4800-94-6 |
|---|---|
| Molecular Formula | C17H16N2Na2O6S |
| Molecular Weight (g/mol) | 422.36 |
| MDL Number | MFCD00077683 |
| InChI Key | RTYJTGSCYUUYAL-YCAHSCEMSA-L |
| Synonym | α-Carboxybenzylpenicillin |
| PubChem CID | 20933 |
| ChEBI | CHEBI:34609 |
| IUPAC Name | disodium;(2S,5R,6R)-6-[(2-carboxylato-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate |
| SMILES | [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN19509201
|
N/A | 1 g |
Each for $182.39
|
|
|||||
|
ICN19509205
|
N/A | 5 g |
Each for $626.88
|
|
|||||
Chemical Identifiers
| 4800-94-6 | |
| 422.36 | |
| RTYJTGSCYUUYAL-YCAHSCEMSA-L | |
| 20933 | |
| disodium;(2S,5R,6R)-6-[(2-carboxylato-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate |
| C17H16N2Na2O6S | |
| MFCD00077683 | |
| α-Carboxybenzylpenicillin | |
| CHEBI:34609 | |
| [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O |
Specifications
| Carbenicillin Disodium Salt | |
| 4800-94-6 | |
| C17H16N2Na2O6S | |
| α-Carboxybenzylpenicillin | |
| [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O | |
| 422.36 | |
| CHEBI:34609 | |
| Carbenicillin disodium salt |
| Powder | |
| Yellow | |
| MFCD00077683 | |
| RTYJTGSCYUUYAL-YCAHSCEMSA-L | |
| disodium;(2S,5R,6R)-6-[(2-carboxylato-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate | |
| 20933 | |
| 422.4 |