missing translation for 'onlineSavingsMsg'
Learn More

Carbenicillin disodium salt, MP Biomedicals™

Catalog No. ICN19509201
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19509201 1 g
ICN19509205 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19509201 Supplier MP Biomedicals Inc Supplier No. 0219509201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4800-94-6 | Formula: C17H16N2Na2O6S | Weight: 422.36 g/mol

Chemical Identifiers

CAS 4800-94-6
Molecular Formula C17H16N2Na2O6S
Molecular Weight (g/mol) 422.36
MDL Number MFCD00077683
InChI Key RTYJTGSCYUUYAL-YCAHSCEMSA-L
Synonym α-Carboxybenzylpenicillin
PubChem CID 20933
ChEBI CHEBI:34609
IUPAC Name disodium;(2S,5R,6R)-6-[(2-carboxylato-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O

Specifications

Type Carbenicillin Disodium Salt
Physical Form Powder
Color Yellow
Quantity 1 g
Formula Weight 422.4
Grade Not Available
Chemical Name or Material Carbenicillin disodium salt