missing translation for 'onlineSavingsMsg'
Learn More

Carbazole-9-carbonyl Chloride 98.0+%, TCI America™

Catalog No. C10311G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10311G 1 g
C10315G 5 g
C103125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C10311G Supplier TCI America Supplier No. C10311G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 73500-82-0
Molecular Formula C13H8ClNO
Molecular Weight (g/mol) 229.663
MDL Number MFCD00059689
InChI Key RCENRFKQKSIRLA-UHFFFAOYSA-N
Synonym 9-(Chlorocarbonyl)carbazole
PubChem CID 3262261
IUPAC Name carbazole-9-carbonyl chloride
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C(=O)Cl

Specifications

Melting Point 105°C
Color White
Quantity 1 g
Formula Weight 229.66
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Carbazole-9-carbonyl Chloride