missing translation for 'onlineSavingsMsg'
Learn More

Carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy, free radical, 99%

CAS: 3229-73-0 | C9H16N2O2 | 184.24 g/mol

$152.53 - $519.00

Chemical Identifiers

CAS 3229-73-0
Molecular Formula C9H16N2O2
Molecular Weight (g/mol) 184.24
MDL Number MFCD00005341
InChI Key GMXOTWPDUBXCMS-UHFFFAOYSA-N
Synonym 2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxamide
IUPAC Name 1-hydroxy-2,2,5,5-tetramethyl-2,5-dihydro-1H-pyrrole-3-carboxamide
SMILES CC1(C)C=C(C(N)=O)C(C)(C)N1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1738103
View Documents
Thermo Scientific Chemicals
A1738103
1 g
Each for $152.53
Only null left
 
AAA1738106
View Documents
Thermo Scientific Chemicals
A1738106
5 g
Each for $519.00
Only null left
 
Description

Description

It is employed as a highly reactive spin-label.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a highly reactive spin-label.

Solubility
Soluble in acetone, chloroform, dichloromethane, ethyl acetate and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

3229-73-0
184.24
GMXOTWPDUBXCMS-UHFFFAOYSA-N
1-hydroxy-2,2,5,5-tetramethyl-2,5-dihydro-1H-pyrrole-3-carboxamide
C9H16N2O2
MFCD00005341
2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxamide
CC1(C)C=C(C(N)=O)C(C)(C)N1O

Specifications

3229-73-0
C9H16N2O2
1 g
2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxamide
GMXOTWPDUBXCMS-UHFFFAOYSA-N
1-hydroxy-2,2,5,5-tetramethyl-2,5-dihydro-1H-pyrrole-3-carboxamide
183.23
Carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy, free radical
198°C to 200°C
MFCD00005341
4138020
Soluble in acetone,chloroform,dichloromethane,ethyl acetate and methanol.
CC1(C)C=C(C(N)=O)C(C)(C)N1O
184.24
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 221-765-9

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only