Learn More
Carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy, free radical, 99%
CAS: 3229-73-0 | C9H16N2O2 | 184.24 g/mol
Supplier: Thermo Scientific Chemicals A1738103
Description
It is employed as a highly reactive spin-label.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a highly reactive spin-label.
Solubility
Soluble in acetone, chloroform, dichloromethane, ethyl acetate and methanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| Carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy | |
| 3229-73-0 | |
| C9H16N2O2 | |
| 1 g | |
| 2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxamide | |
| GMXOTWPDUBXCMS-UHFFFAOYSA-N | |
| 1-hydroxy-2,2,5,5-tetramethyl-2,5-dihydro-1H-pyrrole-3-carboxamide | |
| 183.23 |
| free radical | |
| 198°C to 200°C | |
| MFCD00005341 | |
| 4138020 | |
| Soluble in acetone,chloroform,dichloromethane,ethyl acetate and methanol. | |
| CC1(C)C=C(C(N)=O)C(C)(C)N1O | |
| 184.24 | |
| 99% |
Chemical Identifiers
| 3229-73-0 | |
| 184.24 | |
| GMXOTWPDUBXCMS-UHFFFAOYSA-N | |
| 1-hydroxy-2,2,5,5-tetramethyl-2,5-dihydro-1H-pyrrole-3-carboxamide |
| C9H16N2O2 | |
| MFCD00005341 | |
| 2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxamide | |
| CC1(C)C=C(C(N)=O)C(C)(C)N1O |
Safety and Handling
EINECSNumber : 221-765-9
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only