missing translation for 'onlineSavingsMsg'
Learn More

Carbamazepine, Analytical Standard, ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013844
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013844 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013844 Supplier Merck Emd Millipore Supplier No. 94496100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 298-46-4
Physical Form Neat
Linear Formula C15H12N2O
MDL Number MFCD00005073
Melting Point 191°C to 192°C
Synonym 5 H-Dibenz[b,f]azepine-5-carboxamide
Quantity 100 mg
Formula Weight 236.27
Percent Purity ≥99.0% (HPLC)
Grade Analytical Standard