missing translation for 'onlineSavingsMsg'
Learn More

Carbamazepine, 98%

Catalog No. p-7042455
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6259014 25 g
AAJ6259022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6259014 Supplier Thermo Scientific Chemicals Supplier No. J6259014
Only null left

CAS: 298-46-4 | C15H12N2O | 236.27 g/mol

Carbamazepine is used as cytochrome P450 3A4 inducing anti-epileptic drug and ligand for the GABA-A receptor benzodiazepine modulatory site. Carbamazepine is typically used for the treatment of seizure disorders and neuropathic pain. It is associated with other medications and used to treat schizophrenia as well as a second line agent in bipolar disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in ethanol, acetone, propylene glycol, chloroform, dimethyl sulfoxide, methanol, dimethyl fluoride,ethylene glycol and monoethyl ether. Slightly soluble in glacial acetic acid. Insoluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 298-46-4
Molecular Formula C15H12N2O
Molecular Weight (g/mol) 236.27
MDL Number MFCD00005073
InChI Key FFGPTBGBLSHEPO-UHFFFAOYSA-N
Synonym carbamazepine, tegretol, 5h-dibenzo b,f azepine-5-carboxamide, carbamazepen, carbazepine, finlepsin, carbamezepine, equetro, tegretal, biston
PubChem CID 2554
ChEBI CHEBI:3387
IUPAC Name 2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaene-2-carboxamide
SMILES NC(=O)N1C2=CC=CC=C2C=CC2=CC=CC=C12

Specifications

Melting Point 189°C to 192°C
Color White
Quantity 25 g
Merck Index 14,1781
Solubility Information Soluble in ethanol,acetone,propylene glycol,chloroform,dimethyl sulfoxide,methanol,dimethyl fluoride,ethylene glycol and monoethyl ether. Slightly soluble in glacial acetic acid. Insoluble in water.
Formula Weight 236.27
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Carbamazepine
Hazard Category H302-H317-H334
Hazard Statement GHS H Statement
H334-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280g-P285-P301+P312-P302+P352-P304+P341-P330-P333+P313-P342+P311-P363-P501c
EINECSNumber 206-062-7
RTECSNumber HN8225000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only