accessibility menu, dialog, popup

UserName

CAS RN 298-46-4

IUPAC Name: 2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaene-2-carboxamide
Synonyms: carbamazepine neurotol biston finlepsin atretol karbamazepin calepsin taro-carbamazepine cr novo-carbamaz carbamazepinum
Molecular Weight (g/mol): 236.27
Molecular Formula: C15H12N2O
InChi Key: FFGPTBGBLSHEPO-UHFFFAOYSA-N
SMILES: NC(=O)N1C2=CC=CC=C2C=CC2=CC=CC=C12
Percent Purity 
  • (2)
  • (1)
  • (5)
Physical Form 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)