missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Carbamazepine, 98%

Catalog No. AC344360050
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC344360050 5 g Glass bottle
AC344360010 1 g Glass bottle
AC344360250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC344360050 Supplier Thermo Scientific Chemicals Supplier No. 344360050
Only null left

Carbamazepine (Carbamazepen), CAS # 298-46-4, is an aromatic ring of the dibenzoazepine class that acts as a sodium channel blocker. | CAS: 298-46-4 | C15H12N2O | 236.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Carbamazepine is an aromatic ring of the dibenzoazepine class that acts as a sodium channel blocker
  • It is a tricyclic antidepressant-like drug that reduces post-tetanic potentiation

Applications

  • Carbamazepine can be used as a sodium channel blocker and as a glutamate-transporter agonist to reduce neural activity and produce inhibitory effects in the brain
  • Carbamazepine is useful in the research of pain, epilepsy, and manic depression

Chemical Identifiers

CAS 298-46-4
Molecular Formula C15H12N2O
Molecular Weight (g/mol) 236.27
MDL Number MFCD00005073
InChI Key FFGPTBGBLSHEPO-UHFFFAOYSA-N
Synonym carbamazepine, tegretol, 5h-dibenzo b,f azepine-5-carboxamide, carbamazepen, carbazepine, finlepsin, carbamezepine, equetro, tegretal, biston
PubChem CID 2554
ChEBI CHEBI:3387
SMILES NC(=O)N1C2=CC=CC=C2C=CC2=CC=CC=C12

Specifications

Melting Point 189°C to 192°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 1783
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,acetone,propylene glycol,dmf,slightly soluble in ethanol,glacial acetic acid
IUPAC Name 2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaene-2-carboxamide
Formula Weight 236.27
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material Carbamazepine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POIS
EINECSNumber 206-062-7
RTECSNumber HN8225000

RUO – Research Use Only