missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbamazepine 10,11-epoxide, ≥98% (HPLC), MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Packaging | Poly Bottle |
| CAS | 36507-30-9 |
| Physical Form | Neat |
| Linear Formula | C15H12N2O2 |
| MDL Number | MFCD00467137 |
| Synonym | 1 A,10 b-Dihydro-6 H-dibenzo(b,f)oxireno[d]azepine-6-carboxamide |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 252.27 |
| Percent Purity | ≥98% (HPLC) |
| Show More |