missing translation for 'onlineSavingsMsg'
Learn More

Carbamazepine 10,11-epoxide, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014038
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014038 5 mg
111014039 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014038 Supplier Merck Emd Millipore Supplier No. C42065MG
Only null left

Carbamazepine 10,11-epoxide is a metabolite of the drug carbamazepine.

Pharmaceutical (small molecule)

Specifications

Packaging Poly Bottle
CAS 36507-30-9
Physical Form Neat
Linear Formula C15H12N2O2
MDL Number MFCD00467137
Synonym 1 A,10 b-Dihydro-6 H-dibenzo(b,f)oxireno[d]azepine-6-carboxamide
Type Analytical Standard
Quantity 5 mg
Formula Weight 252.27
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less